Ethyl-3-brom-6-chlorimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: Apollo Scientific

OR110198-1G OR110198-5G
APOSOR110198-1GEA 102 EUR
APOSOR110198-1G APOSOR110198-5G
Ethyl-3-brom-6-chlorimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-3-brom-6-chlorimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₀H₈BrClN₂O₂
Schmelzpunkt: 139…141 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06496148
CAS-Nummer: 861208-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR