Dimethyl hydrogen phosphate
Lieferant: Apollo Scientific
|
Gefahr
|
OR10807-1G
OR10807-5G
APOSOR10807-1GEA
65.4
EUR
APOSOR10807-1G
APOSOR10807-5G
Dimethyl hydrogen phosphate
Dimethyl hydrogen phosphate
Formel:
C₂H₇O₄P Siedepunkt: 174 °C Dichte: 1.323 g/cm³ (25 °C) Lagerungstemperatur: Raumtemperatur |
CAS-Nummer:
813-78-5 EINECS: 212-389-6 UN-Nummer: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...