4-(1-Pyrrolidinylcarbonyl)phenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(1-Pyrrolidinylcarbonyl)benzeneboronic acid, 4-(Pyrrolidine-1-carbonyl)phenylboronic acid, (4-(Pyrrolidine-1-carbonyl)phenyl)boronic acid
OR10584-1G
OR10584-5G
APOSOR10584-1GEA
98.6
EUR
APOSOR10584-1G
APOSOR10584-5G
4-(1-Pyrrolidinylcarbonyl)phenylboronsäure
4-(1-Pyrrolidinylcarbonyl)phenylboronsäure
Formel:
C₁₁H₁₄BNO₃ Schmelzpunkt: 136…140 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD03411950 CAS-Nummer: 389621-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...