E-Phenylethenylboronic acid 95%

Lieferant: Apollo Scientific
Achtung

OR10578-5G OR10578-25G
APOSOR10578-5GEA 129 EUR
APOSOR10578-5G APOSOR10578-25G
E-Phenylethenylboronic acid 95%
E-Phenylethenylboronic acid
Formel: C₈H₉BO₂
Schmelzpunkt: 146…156 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00963621
CAS-Nummer: 6783-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR