2-Carbamoylbenzeneboronic acid 98%

Lieferant: Apollo Scientific
Achtung

OR10324-1G OR10324-5G
APOSOR10324-1GEA 66.7 EUR
APOSOR10324-1G APOSOR10324-5G
2-Carbamoylbenzeneboronic acid 98%
2-Carbamoylbenzeneboronic acid
Formel: C₇H₈BNO₃
Schmelzpunkt: 226…230 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD02179454
CAS-Nummer: 380430-54-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR