ɑ-Cyclohexylphenylacetic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: ['2-Cyclohexyl-2-phenylacetic acid'], α-Cyclohexylphenylacetic acid, 2-Cyclohexyl-2-phenylacetic Acid

OR0997-25G OR0997-5G
APOSOR0997-25GEA 203 EUR
APOSOR0997-25G APOSOR0997-5G
ɑ-Cyclohexylphenylacetic acid 95%
alpha-Cyclohexylphenylacetic acid
Formel: C₁₄H₁₈O₂
Schmelzpunkt: 148…152 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00001477
CAS-Nummer: 3894-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR