1-(Boc-amino)cyclobutanecarboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-BOC-1-Aminocyclobutanecarboxylic acid, Boc-cyclovaline, N-tert-Butoxycarbonyl-1-aminocyclobutanecarboxylic acid, 1-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid, N-Boc-1-aminocyclobutanecarboxylic acid

OR0918-100G OR0918-25G
APOSOR0918-100GEA 202 EUR
APOSOR0918-100G APOSOR0918-25G
1-(Boc-amino)cyclobutanecarboxylic acid 97%
1-(Boc-amino)cyclobutanecarboxylic acid
Formel: C₁₀H₁₇NO₄
Schmelzpunkt: 129…134 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD02682623
CAS-Nummer: 120728-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR