4-(Benzyloxy)-3-chlorbenzaldehyd 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR018832-1G
OR018832-5G
APOSOR018832-1GEA
54.4
EUR
APOSOR018832-1G
APOSOR018832-5G
4-(Benzyloxy)-3-chlorbenzaldehyd 95+%
4-(Benzyloxy)-3-chlorbenzaldehyd
Formel:
C₁₄H₁₁ClO₂ Schmelzpunkt: 70…71 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00831645 CAS-Nummer: 66422-84-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...