2-Carbomethoxybenzylamine hydrochloride 95+%

Lieferant: Apollo Scientific
Achtung

OR0170-1G OR0170-5G OR0170-10G
APOSOR0170-1GEA 0 EUR
APOSOR0170-1G APOSOR0170-5G APOSOR0170-10G
2-Carbomethoxybenzylamine hydrochloride 95+%
2-Carbomethoxybenzylamine hydrochloride
Formel: C₉H₁₂ClNO₂
Schmelzpunkt: 170…172 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06245496
CAS-Nummer: 849020-92-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR