(2-Aminoethyl)benzyl carbamic acid-tert-butylester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Benzylethane-1,2-diamine, N-BOC protected
OR01637-1G
OR01637-5G
APOSOR01637-1GEA
52.2
EUR
APOSOR01637-1G
APOSOR01637-5G
(2-Aminoethyl)benzyl carbamic acid-tert-butylester
(2-Aminoethyl)benzyl carbamic acid-tert-butylester
Formel:
C₁₄H₂₂N₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06245501 CAS-Nummer: 152193-00-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...