Thuliumtrinitrat Pentahydrat 99,9%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Thulium trinitrate pentahydrate
IN3607-1G
IN3607-5G
APOSIN3607-1GEA
102
EUR
APOSIN3607-1G
APOSIN3607-5G
Thuliumtrinitrat Pentahydrat 99,9%
Thuliumtrinitrat Pentahydrat
Formel:
N₃O₉Tm·5H₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149875 CAS-Nummer: 36548-87-5 UN-Nummer: 1477 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...