meta-Topolin

Lieferant: Apollo Scientific
Achtung

BIT0841-100MG BIT0841-1G BIT0841-500MG
APOSBIT0841-100MGEA 50.9 EUR
APOSBIT0841-100MG APOSBIT0841-1G APOSBIT0841-500MG
meta-Topolin
meta-Topolin

Highly active aromatic cytokinin from poplar leaves.

Formel: C₁₂H₁₁N₅O
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD03065993
CAS-Nummer: 75737-38-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR