Boceprevir
Lieferant: Apollo Scientific
BISN0326-5MG
BISN0326-50MG
BISN0326-100MG
APOSBISN0326-5MGEA
92.8
EUR
APOSBISN0326-5MG
APOSBISN0326-50MG
APOSBISN0326-100MG
Boceprevir
Boceprevir
Formel:
C₂₇H₄₅N₅O₅ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD22208555 CAS-Nummer: 394730-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...