B-Raf inhibitor 1
Lieferant: Apollo Scientific
Synonyme:
N5-(3-(7H-purin-6-yl)pyridin-2-yl)-N1-(4-chlorophenyl)-6-methylisoquinoline-1,5-diamine
BISN0223-1MG
BISN0223-5MG
BISN0223-10MG
APOSBISN0223-1MGEA
79.9
EUR
APOSBISN0223-1MG
APOSBISN0223-5MG
APOSBISN0223-10MG
B-Raf inhibitor 1
B-Raf inhibitor 1
MAPK/ERK Signalling Pathway
Formel:
C₂₆H₁₉ClN₈ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD22571725 CAS-Nummer: 1093100-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...