FABP4 inhibitor

Lieferant: Apollo Scientific

Synonyme: BMS-309403, 2-((2'-(5-Ethyl-3,4-diphenyl-1H-pyrazol-1-yl)-[1,1'-biphenyl]-3-yl)oxy)acetic acid

BISN0155-5MG BISN0155-25MG
APOSBISN0155-5MGEA 132 EUR
APOSBISN0155-5MG APOSBISN0155-25MG
FABP4 inhibitor
FABP4 inhibitor

BMS-309403 is an inhibitor of fatty acid binding protein 4 FABP4 with Ki of less than 2 nM that decreases fatty acid uptake in adipocytes in vitro.

Formel: C₃₁H₂₆N₂O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09991687
CAS-Nummer: 300657-03-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR