Spectinomycin dihydrochloride pentahydrate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-4a,7,9-Trihydroxy-2-methyl-6,8-bis(methylamino)decahydro-4H-benzo[b]pyrano[2,3-e][1,4]dioxin-4-one dihydrochloride pentahydrate
BIS0147-25G
BIS0147-5G
APOSBIS0147-25GEA
85.7
EUR
APOSBIS0147-25G
APOSBIS0147-5G
Spectinomycin dihydrochloride pentahydrate
Spectinomycin dihydrochloride pentahydrate
An aminocyclitol antibiotic which acts by binding to the 30S subunit of the bacterial ribosome and inhibiting protein synthesis.
- Cell culture tested
Formel:
C₁₄H₂₄N₂O₇·2HCl·5H₂O Schmelzpunkt: 210 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00150886 CAS-Nummer: 22189-32-8 EINECS: 244-554-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...