Boc-Amino-PEG3-Amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl (2-(2-(2-(2-aminoethoxy)ethoxy)ethoxy)ethyl)carbamate, NH2-PEG3-C2-NH-Boc
BIPG1811-100MG
BIPG1811-50MG
BIPG1811-250MG
APOSBIPG1811-100MGEA
125
EUR
APOSBIPG1811-100MG
APOSBIPG1811-50MG
APOSBIPG1811-250MG
Boc-Amino-PEG3-Amine
Boc-Amino-PEG3-Amine
Formel:
C₁₃H₂₈N₂O₅ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD16619220 CAS-Nummer: 101187-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...