t-Boc-N-amido-PEG8-acid

Lieferant: Apollo Scientific

BIPG1805-50MG BIPG1805-100MG
APOSBIPG1805-50MGEA 68.3 EUR
APOSBIPG1805-50MG APOSBIPG1805-100MG
t-Boc-N-amido-PEG8-acid
t-Boc-N-amido-PEG8-acid
Formel: C₂₄H₄₇NO₁₂
Lagerungstemperatur: Kühlschrank
CAS-Nummer: 1334169-93-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR