Mal-amido-PEG4-acid

Lieferant: Apollo Scientific

Synonyme: 19-Maleimido-17-oxo-4,7,10,13-tetraoxa-16-azanonadecanoic acid, 1-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13,16-tetraoxa-4-azanonadecan-19-oic acid

BIPG1473-1G BIPG1473-250MG
APOSBIPG1473-1GEA 171 EUR
APOSBIPG1473-1G APOSBIPG1473-250MG
Mal-amido-PEG4-acid
Mal-amido-PEG4-acid
Formel: C₁₈H₂₈N₂O₉
Molekulargewicht: 416.42 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD13184954
CAS-Nummer: 1263045-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR