(4-Nitrophenyl)-β-D-galactopyranosid
Lieferant: Apollo Scientific
BIN0431-10G
BIN0431-5G
APOSBIN0431-10GEA
364
EUR
APOSBIN0431-10G
APOSBIN0431-5G
(4-Nitrophenyl)-β-D-galactopyranosid
(4-Nitrophenyl)-β-D-galactopyranosid
Formel:
C₁₂H₁₅NO₈ Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00063256 CAS-Nummer: 3150-24-1 EINECS: 221-584-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...