2'-Fluoro-2'-deoxyadenosine

Lieferant: Apollo Scientific
Achtung

BIMN2001-1G BIMN2001-500MG BIMN2001-5G
APOSBIMN2001-1GEA 178 EUR
APOSBIMN2001-1G APOSBIMN2001-500MG APOSBIMN2001-5G
2'-Fluoro-2'-deoxyadenosine
2'-Fluoro-2'-deoxyadenosine
Formel: C₁₀H₁₂FN₅O₃
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09750859
CAS-Nummer: 64183-27-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR