H-89 dihydrochloride hydrate ≥99%

Lieferant: Apollo Scientific
Achtung

BIH127-10MG BIH127-25MG BIH127-5MG
APOSBIH127-10MGEA 68.3 EUR
APOSBIH127-10MG APOSBIH127-25MG APOSBIH127-5MG
H-89 dihydrochloride hydrate ≥99%
H-89 dihydrochloride hydrate

Potent and selective inhibitor of PKA, with an IC50 of about 50 nM.  J. Biol. Chem. 265:  5267-5272 (1990)


  • Protein

Formel: C₂₀H₂₀BrN₃O₂S·2HCl
Lagerungstemperatur: Gefrierschrank
CAS-Nummer: 130964-39-5 | 127243-85-0 (free base)

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR