D(+)-Glucose 6-phosphate sodium salt
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
ROBISON ESTER MONOSODIUM SALT, Sodium (2R,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexyl hydrogenphosphate, G-6-P Na, BETA-D-GLUCOSE 6-PHOSPHATE MONOSODIUM SALT, D-Glucose 6-phosphate sodium
BIG5011-10G
BIG5011-1G
APOSBIG5011-10GEA
589
EUR
APOSBIG5011-10G
APOSBIG5011-1G
D(+)-Glucose 6-phosphate sodium salt
D-(+)-Glucose-6-phosphate sodium salt
Stable alternative to disodium salt.
Formel:
C₆H₁₂NaO₉P Schmelzpunkt: 204 °C (dec.) Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00006611 CAS-Nummer: 54010-71-8 EINECS: 258-921-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...