Geldanamycin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4E,6Z,8S,9S,10E,12S,13R,14S,16R)-13-Hydroxy-8,14,19-trimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate
BIG2461-100MG
BIG2461-250MG
BIG2461-25MG
APOSBIG2461-100MGEA
284
EUR
APOSBIG2461-100MG
APOSBIG2461-250MG
APOSBIG2461-25MG
Geldanamycin
Geldanamycin
Potent antitumour antibiotic.
Formel:
C₂₉H₄₀N₂O₉ Molekulargewicht: 560.64 g/mol Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00274570 CAS-Nummer: 30562-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...