2'-Deoxyguanosine hydrate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
9-(2-Deoxy-β-D-ribofuranosyl)guanine hydrate, Guanine-2′-deoxyriboside hydrate, 2-Amino-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one hydrate(1:x)
BIG1008-1G
BIG1008-500MG
BIG1008-5G
APOSBIG1008-1GEA
29
EUR
APOSBIG1008-1G
APOSBIG1008-500MG
APOSBIG1008-5G
2'-Deoxyguanosine hydrate
2'-Deoxyguanosine hydrate
Formel:
C₁₀H₁₃N₅O₄·xH₂O Schmelzpunkt: 300 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00150760 CAS-Nummer: 961-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...