D-Glucoronic acid
Lieferant: Apollo Scientific
|
Achtung
|
BIG1006-100G
BIG1006-25G
BIG1006-50G
APOSBIG1006-100GEA
299
EUR
APOSBIG1006-100G
APOSBIG1006-25G
APOSBIG1006-50G
D-Glucoronic acid
D-Glucoronic acid
Formel:
C₆H₁₀O₇ Schmelzpunkt: 158 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00166981 CAS-Nummer: 6556-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...