L(-)-Glucose 98%

Lieferant: Apollo Scientific
Achtung

BIFS0106-1G BIFS0106-5G BIFS0106-10G
APOSBIFS0106-1GEA 66.7 EUR
APOSBIFS0106-1G APOSBIFS0106-5G APOSBIFS0106-10G
L(-)-Glucose 98%
L(-)-Glucose
Formel: C₆H₁₂O₆
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00148913
CAS-Nummer: 921-60-8
EINECS: 213-068-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR