Ruxolitinib freie Base ≥99%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(R)-3-(4-(7H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl)-3-cyclopentylpropanenitrile, INCB018424, Ruxolitinib
BIFK0027-10MG
BIFK0027-25MG
BIFK0027-5MG
APOSBIFK0027-10MGEA
87
EUR
APOSBIFK0027-10MG
APOSBIFK0027-25MG
APOSBIFK0027-5MG
Ruxolitinib freie Base ≥99%
Ruxolitinib
Formel:
C₁₇H₁₈N₆ Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD12031592 CAS-Nummer: 941678-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...