Fluorescein di-β-D-glucopyranoside

Lieferant: Apollo Scientific

BICS0116-2MG BICS0116-5MG BICS0116-1MG
APOSBICS0116-2MGEA 596 EUR
APOSBICS0116-2MG APOSBICS0116-5MG APOSBICS0116-1MG
Fluorescein di-β-D-glucopyranoside
Fluorescein di-β-D-glucopyranoside
Formel: C₃₂H₃₂O₁₅
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 129787-66-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR