6-Azido-6-deoxy-D-galactopyranose 98% min

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Azido-6-deoxy-D-galactose, (2R,3S,4S,5R)-6-Azido-2,3,4,5-tetrahydroxyhexanal

BICL4317-1G BICL4317-250MG BICL4317-500MG
APOSBICL4317-1GEA 1210 EUR
APOSBICL4317-1G APOSBICL4317-250MG APOSBICL4317-500MG
6-Azido-6-deoxy-D-galactopyranose 98% min
6-Azido-6-deoxy-D-galactopyranose
Formel: C₆H₁₁N₃O₅
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD03265529
CAS-Nummer: 66927-03-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR