4-Methylphenyl-β-D-thiogalactopyranoside 99% min
Lieferant: Apollo Scientific
|
Achtung
|
BICL4167-10G
BICL4167-5G
BICL4167-2G
APOSBICL4167-10GEA
824
EUR
APOSBICL4167-10G
APOSBICL4167-5G
APOSBICL4167-2G
4-Methylphenyl-β-D-thiogalactopyranoside 99% min
4-Methylphenyl-beta-D-thiogalactopyranoside
Formel:
C₁₃H₁₈O₅S Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD04973631 CAS-Nummer: 28244-98-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...