MBS (N-Succinimidyl 3-maleimidobenzoate)
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Maleimidobenzoic acid N-hydroxysuccinimide ester, 3-Maleimidobenzoic Acid N-Succinimidyl Ester, MBS, m-Maleimidobenzoyl-N-hydroxysuccinimide ester, N-(3-Maleimidobenzoyloxy)succinimide, 2,5-Dioxopyrrolidin-1-yl 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoate
Primary amine reactive -Sulphhydryl reactive. Protein conjugation Hapten carrier Protein conjugation
Formel:
C₁₅H₁₀N₂O₆ Schmelzpunkt: 175…177 °C Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00005514 CAS-Nummer: 58626-38-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...