Cefoperazone sodium salt
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Cefoneg, Cefosint
BIC1033-25G
BIC1033-5G
APOSBIC1033-25GEA
270
EUR
APOSBIC1033-25G
APOSBIC1033-5G
Cefoperazone sodium salt
Cefoperazone sodium salt
Formel:
C₂₅H₂₆N₉NaO₈S₂ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD07793331 CAS-Nummer: 62893-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...