Fmoc-δ-azido-Nva-OH ≥98%

Lieferant: Apollo Scientific

Synonyme: Fmoc-Orn(N3)

BIBP1083-250MG BIBP1083-1G BIBP1083-5G
APOSBIBP1083-250MGEA 130 EUR
APOSBIBP1083-250MG APOSBIBP1083-1G APOSBIBP1083-5G
Fmoc-δ-azido-Nva-OH ≥98%
Fmoc-δ-azido-Nva-OH
Formel: C₂₀H₂₀N₄O₄
Molekulargewicht: 380.40 g/mol
Lagerungstemperatur: Kühlschrank
CAS-Nummer: 1097192-04-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR