4-Methylumbelliferyl sulphate potassium salt

Lieferant: Apollo Scientific
Achtung

Synonyme: Potassium 4-methyl-2-oxo-2H-chromen-7-yl sulphate, 4-Methylumbelliferyl sulphate potassium

BIB6119-1G BIB6119-5G
APOSBIB6119-1GEA 79.9 EUR
APOSBIB6119-1G APOSBIB6119-5G
4-Methylumbelliferyl sulphate potassium salt
4-Methylumbelliferyl sulphate potassium salt
Formel: C₁₀H₇KO₆S
Molekulargewicht: 294.318 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD00016970
CAS-Nummer: 15220-11-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR