DL-Methioninsulfoxid

Lieferant: Apollo Scientific

Synonyme: 2-Amino-4-(methylsulphinyl)butanoic acid

BIB6099-10G BIB6099-5G
APOSBIB6099-10GEA 109 EUR
APOSBIB6099-10G APOSBIB6099-5G
DL-Methioninsulfoxid
DL-Methioninsulfoxid
Formel: C₅H₁₁NO₃S
Schmelzpunkt: 240 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00002620
CAS-Nummer: 62697-73-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR