L-Histidinol dihydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
L-Histidinol·2 HCl, (S)-2-Amino-3-(1H-imidazol-4-yl)propan-1-ol dihydrochloride
BIB6064-10G
BIB6064-5G
APOSBIB6064-10GEA
319
EUR
APOSBIB6064-10G
APOSBIB6064-5G
L-Histidinol dihydrochloride
L-Histidinol dihydrochloride
Formel:
C₆H₁₃Cl₂N₃O Schmelzpunkt: 198…201 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00078058 CAS-Nummer: 1596-64-1 EINECS: 216-482-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...