Gly pro-7-amido-4-methylcoumarin hydrobromide

Lieferant: Apollo Scientific
Achtung

Synonyme: H-Gly-Pro-AMC·HBr

BIB6062-100MG BIB6062-25MG
APOSBIB6062-100MGEA 357 EUR
APOSBIB6062-100MG APOSBIB6062-25MG
Gly pro-7-amido-4-methylcoumarin hydrobromide
Gly pro-7-amido-4-methylcoumarin hydrobromide

Substratefor dipeptidylaminopeptidase IV.

Formel: C₁₇H₂₀BrN₃O₄
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD00152092
CAS-Nummer: 115035-46-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR