D(+)-Cellobiose

Lieferant: Apollo Scientific
Achtung

BIB6025-100G BIB6025-25G
APOSBIB6025-100GEA 114 EUR
APOSBIB6025-100G APOSBIB6025-25G
D(+)-Cellobiose
D(+)-Cellobiose
Formel: C₁₂H₂₂O₁₁
Schmelzpunkt: 239…240 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00136034
CAS-Nummer: 528-50-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR