Bicin ≥99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine
BIB1151-100G
BIB1151-1KG
BIB1151-2.5KG
APOSBIB1151-100GEA
21.9
EUR
APOSBIB1151-100G
APOSBIB1151-1KG
APOSBIB1151-2.5KG
Bicin ≥99%
Bicin
Biological buffer - useful range 7.6 - 9.0. Recommended for low temperature work.
Formel:
C₆H₁₃NO₄ Schmelzpunkt: 190 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00004295 CAS-Nummer: 150-25-4 EINECS: 205-755-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...