Tricain-Methansulfonat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
TS 222, 3-Aminobenzoic acid Ethyl Ester Methanesulphonate, Tricaine, Ethyl 3-aminobenzoate methanesulphonate, Ethyl 3-aminobenzoate, methanesulphonic acid salt, Bio-Calm™, Ethyl-m-Aminobenzoate, Methanesulphonate salt, MS-222
BIA1347-25G
BIA1347-50G
BIA1347-100G
APOSBIA1347-25GEA
43.5
EUR
APOSBIA1347-25G
APOSBIA1347-50G
APOSBIA1347-100G
Tricain-Methansulfonat 98%
Tricain-Methansulfonat
Formel:
C₁₀H₁₅NO₅S Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00013176 CAS-Nummer: 886-86-2 EINECS: 212-956-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...