N-(2-Acetamido)-2-aminoethansulfonsäure (ACES) ≥99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid
BIA0301-100G
BIA0301-250G
BIA0301-500G
APOSBIA0301-100GEA
36.4
EUR
APOSBIA0301-100G
APOSBIA0301-250G
APOSBIA0301-500G
N-(2-Acetamido)-2-aminoethansulfonsäure (ACES) ≥99%
N-(2-Acetamido)-2-aminoethansulfonsäure (ACES)
An efficient separator in the resolution of proteinsystems by IEF. Biological buffer - useful range 6.4 - 7.4
Formel:
H₂NCOCH₂NHCH₂CH₂SO₃H Molekulargewicht: 182.2 g/mol Schmelzpunkt: 270…295 °C (Zers.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00008030 CAS-Nummer: 7365-82-4 EINECS: 230-908-4 Merck Index: 13,00037 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...