Chlortetracyclin Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
BI7654-10G
BI7654-100G
BI7654-25G
APOSBI7654-10GEA
50.9
EUR
APOSBI7654-10G
APOSBI7654-100G
APOSBI7654-25G
Chlortetracyclin Hydrochlorid
Chlortetracyclin Hydrochlorid
Formel:
C₂₂H₂₃ClN₂O₈·HCl Schmelzpunkt: 210 °C Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD26142699 CAS-Nummer: 64-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...