2,6-Difluoro-4-hydrazinopyridine 97%

Lieferant: Apollo Scientific
Gefahr

PC9683-100MG PC9683-1G PC9683-250MG
APOSPC9683-100MGEA 34.8 EUR
APOSPC9683-100MG APOSPC9683-1G APOSPC9683-250MG
2,6-Difluoro-4-hydrazinopyridine 97%
2,6-Difluoro-4-hydrazinopyridine
Formel: C₅H₅F₂N₃
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD08436095
CAS-Nummer: 837364-94-0
UN-Nummer: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR