Ethyl 2-[3-(trifluoromethyl)phenyl]glyoxylate, tech

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-oxo-2-(3-(trifluoromethyl)phenyl)acetate

PC9274-1G PC9274-5G
APOSPC9274-1GEA 230 EUR
APOSPC9274-1G APOSPC9274-5G
Ethyl 2-[3-(trifluoromethyl)phenyl]glyoxylate, tech
Ethyl 2-[3-(trifluoromethyl)phenyl]glyoxylate
Formel: C₁₁H₉F₃O₃
Siedepunkt: 66…72 °C (0.2 mm Hg)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01934868
CAS-Nummer: 110193-60-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR