2-Chloro-6-fluoro-4-(trifluoromethyl)phenylhydrazine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-5-fluoro-4-hydrazinobenzotrifluoride
PC9168-1G
PC9168-250MG
APOSPC9168-1GEA
193
EUR
APOSPC9168-1G
APOSPC9168-250MG
2-Chloro-6-fluoro-4-(trifluoromethyl)phenylhydrazine 98%
2-Chloro-6-fluoro-4-(trifluoromethyl)phenylhydrazine
Formel:
C₇H₅ClF₄N₂ Schmelzpunkt: 73…75 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD04972729 CAS-Nummer: 110499-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...