2-Bromo-3,6-difluoropyridine 95%
Lieferant: Apollo Scientific
PC911877-100MG
PC911877-1G
PC911877-250MG
APOSPC911877-100MGEA
364
EUR
APOSPC911877-100MG
APOSPC911877-1G
APOSPC911877-250MG
2-Bromo-3,6-difluoropyridine 95%
2-Bromo-3,6-difluoropyridine
Formel:
C₅H₂BrF₂N Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD27939127 CAS-Nummer: 1382786-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...