(2,2,2-Trifluoroethoxy)acetic acid 98%

Lieferant: Apollo Scientific

PC911111-1G PC911111-5G
APOSPC911111-1GEA 197 EUR
APOSPC911111-1G APOSPC911111-5G
(2,2,2-Trifluoroethoxy)acetic acid 98%
(2,2,2-Trifluoroethoxy)acetic acid
Formel: C₄H₅F₃O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02322998
CAS-Nummer: 675-67-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR