(3S)-3-Amino-4-(4-fluorophenyl)butanoic acid hydrochloride 98%
Lieferant: Apollo Scientific
Synonyme:
(S)-3-Amino-4-(4-fluorophenyl)butanoic acid hydrochloride
PC911010-1G
PC911010-250MG
APOSPC911010-1GEA
377
EUR
APOSPC911010-1G
APOSPC911010-250MG
(3S)-3-Amino-4-(4-fluorophenyl)butanoic acid hydrochloride 98%
(3S)-3-Amino-4-(4-fluorophenyl)butanoic acid hydrochloride
Formel:
C₁₀H₁₂FNO₂·ClH Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01861012 CAS-Nummer: 331763-68-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...