6-Fluoro-2-methylbenzo[d]thiazole 98%

Lieferant: Apollo Scientific

PC910716-1G PC910716-250MG PC910716-5G
APOSPC910716-1GEA 169 EUR
APOSPC910716-1G APOSPC910716-250MG APOSPC910716-5G
6-Fluoro-2-methylbenzo[d]thiazole 98%
6-Fluoro-2-methylbenzo[d]thiazole
Formel: C₈H₆FNS
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00012319
CAS-Nummer: 399-73-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR